Learn More
Aviva Systems Biology Corp TEAD1 Antibody - N-terminal region (P100891_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp P100891_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TEAD1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TEAD1. Alternate names: AA, REF1, TCF13, TEF-1, NTEF-1, TCF-13, TEAD-1ApplicationWB. Species reactivity: Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62767