Learn More
Aviva Systems Biology Corp TEAD1 Antibody - C-terminal region (ARP39521_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39521_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TEAD1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human TEAD1. Alternate names: AA, REF1, TCF13, TEF-1, NTEF-1, TCF-13, TEAD-1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57071