Learn More
Aviva Systems Biology Corp TEAD1 Antibody - middle region (ARP31239_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31239_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TEAD1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TEAD1. Alternate names: AA, REF1, TCF13, TEF-1, NTEF-1, TCF-13, TEAD-1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54188