Learn More
ABclonal Technology TEAD2 Rabbit pAb
SDP
Supplier:  ABclonal Technology A155941000UL
Encompass_Preferred
Enables several functions, including DNA-binding transcription factor activity, disordered domain specific binding activity, and transcription coactivator binding activity. Involved in hippo signaling, positive regulation of transcription, DNA-templated, and protein-containing complex assembly. Located in cytosol and nucleoplasm. Part of TEAD-YAP complex.
Catalog No. 50-001-05895