Learn More
Aviva Systems Biology Corp TEAD3 Antibody - C-terminal region (ARP38278_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38278_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TEAD3. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human TEAD3. Alternate names: TEF5, TEAD5, TEF-5, DTEF-1, ETFR-1, TEAD-3ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56591