Learn More
Aviva Systems Biology Corp TEAD3 Antibody - middle region (ARP34065_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34065_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TEAD3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TEAD3. Alternate names: DTEF-1, ETFR-1, TEAD5, TEF-5, TEF5, TEAD-3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55236