Learn More
Medchemexpress LLC TEAD3 Human His-SU 10ug
SDP
Catalog No. 5000196319
Encompass_Preferred
Recombinant human TEAD3 protein fragment (amino acids M112-D435) produced in E. coli and supplied with N-terminal SUMO and 6xHis tags for purification and detection. The product has an approximate molecular weight of 52.3 kDa and a reported purity of about 90%, provided in small research quantities suitable for biochemical and binding assays.
  • N-terminal SUMO and 6xHis tags for affinity purification.
  • Fragment corresponds to amino acids M112-D435 of human TEAD3.
  • Approximate molecular weight 52.3 kDa.
  • Purity around 90% by supplier specification.
  • Available in small quantities for biochemical and binding studies.
Catalog No. 50-001-96319 Supplier Medchemexpress LLC Supplier No. HYP7162610UG
Recombinant human TEAD3 protein fragment (amino acids M112-D435) produced in E. coli and supplied with N-terminal SUMO and 6xHis tags for purification and detection. The product has an approximate molecular weight of 52.3 kDa and a reported purity of about 90%, provided in small research quantities suitable for biochemical and binding assays.
  • N-terminal SUMO and 6xHis tags for affinity purification.
  • Fragment corresponds to amino acids M112-D435 of human TEAD3.
  • Approximate molecular weight 52.3 kDa.
  • Purity around 90% by supplier specification.
  • Available in small quantities for biochemical and binding studies.