Learn More
Medchemexpress LLC TEAD3 Human His-SU 50ug
SDP
Catalog No. 5000195388
Encompass_Preferred
Recombinant human TEAD3 (residues M112-D435) is supplied as an N-terminal SUMO-6xHis tagged protein expressed in E. coli. The purified protein (~52.3 kDa) is provided at 90% purity for biochemical and structural studies and is suitable for binding assays, structural characterization, and in vitro functional assays.
  • Expressed in E. coli
  • N-terminal SUMO and 6xHis affinity tags
  • Approximately 52.3 kDa molecular weight
  • Purity approximately 90%
  • Includes residues M112-D435 of the human sequence
  • Supplied in microgram pack sizes for laboratory use
Catalog No. 50-001-95388 Supplier Medchemexpress LLC Supplier No. HYP7162650UG
Recombinant human TEAD3 (residues M112-D435) is supplied as an N-terminal SUMO-6xHis tagged protein expressed in E. coli. The purified protein (~52.3 kDa) is provided at 90% purity for biochemical and structural studies and is suitable for binding assays, structural characterization, and in vitro functional assays.
  • Expressed in E. coli
  • N-terminal SUMO and 6xHis affinity tags
  • Approximately 52.3 kDa molecular weight
  • Purity approximately 90%
  • Includes residues M112-D435 of the human sequence
  • Supplied in microgram pack sizes for laboratory use