Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC TEAD3 Human His-SU 5ug
SDP

Catalog No. 5000196043
Encompass_Preferred

Recombinant human TEAD3 protein (residues M112-D435) with an N-terminal 6xHis-SUMO fusion tag, supplied as a small-scale purified protein for research applications.

  • Contains residues M112-D435 of human TEAD3.
  • N-terminal 6xHis-SUMO tag for affinity purification.
  • Approximate molecular weight 52.3 kDa.
  • Purity ~90% suitable for biochemical assays.
  • Provided in a 5 μg size for small-scale experiments.

Catalog No. 50-001-96043 Supplier Medchemexpress LLC Supplier No. HYP716265UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human TEAD3 protein (residues M112-D435) with an N-terminal 6xHis-SUMO fusion tag, supplied as a small-scale purified protein for research applications.

  • Contains residues M112-D435 of human TEAD3.
  • N-terminal 6xHis-SUMO tag for affinity purification.
  • Approximate molecular weight 52.3 kDa.
  • Purity ~90% suitable for biochemical assays.
  • Provided in a 5 μg size for small-scale experiments.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.