Learn More
Thomas Scientific TEAR-AWAY PCR PLATE,CLR,8STRIP
SDP
Catalog No. NC1461540
Encompass_Preferred
96 Well 8-strip Tear-A-Way PCR Plate, Clear.Tear-A-Way™ plates allow for the most flexible and efficient use of a PCR plate. No need to run half-empty plates or to cut plates with scissors. Cutting plates risks perforating wells, damaging sealing rings and contamination.Tear-A-Way™ plates can be quickly and easily separated along the perforations between the rows. Every single strip can be utilized, saving time and costs. Please see our website to view the compatibility of Tear-A-Way™ with your thermal cycler or sequencer.Plate is perforated to enable accurate tearing into stripsNumbers on all strips to prevent sample errorsNon-skirted universal cycler and sequencer compatibilitySealable with cap strips, adhesive seals and heat sealsSimply snaps to fit a 24 or 48-well thermal cycler block
Catalog No. NC1461540 Supplier Thomas Scientific Supplier No. 1149U71
Add to Cart
Add to Cart
96 Well 8-strip Tear-A-Way PCR Plate, Clear.Tear-A-Way™ plates allow for the most flexible and efficient use of a PCR plate. No need to run half-empty plates or to cut plates with scissors. Cutting plates risks perforating wells, damaging sealing rings and contamination.Tear-A-Way™ plates can be quickly and easily separated along the perforations between the rows. Every single strip can be utilized, saving time and costs. Please see our website to view the compatibility of Tear-A-Way™ with your thermal cycler or sequencer.Plate is perforated to enable accurate tearing into stripsNumbers on all strips to prevent sample errorsNon-skirted universal cycler and sequencer compatibilitySealable with cap strips, adhesive seals and heat sealsSimply snaps to fit a 24 or 48-well thermal cycler block