Learn More
Cole-Parmer Argos Technologies Teasing Needle, 6", Straight, 19 Guage, White; Each
Catalog No. 501960661 Shop All Cole-Parmer Products
Encompass_Preferred
Needles are made from high quality nichrome wire that cools rapidly after sterilization. Available with or without handle and straight or angled needle.
Catalog No. 50-196-0661 Supplier Cole-Parmer Supplier No. 0439647
Needles are made from high quality nichrome wire that cools rapidly after sterilization. Available with or without handle and straight or angled needle.