Learn More
Medchemexpress LLC Tebanicline dihydrochloride | 209326-19-2 | 99.5% | C9H13Cl3N2O | 1 ML
SDP
Catalog No. 5000354772
Encompass_Preferred
Tebanicline dihydrochloride is a potent, orally effective analgesic that modulates neuronal nicotinic acetylcholine receptors (nAChRs). It exhibits high selectivity for neuronal nAChRs, acting as an agonist, and effectively inhibits cytisine binding to α4β2 neuronal nAChRs. This product is investigated for its potential in treating various types of pain and for its anxiolytic-like properties.
  • Potent and orally effective analgesic.
  • Highly selective for neuronal nAChRs.
  • Shows efficacy in models of acute, persistent, chemical, and neuropathic pain.
  • Investigated for anxiolytic-like activity.
Catalog No. 50-003-54772 Supplier Medchemexpress LLC Supplier No. HY14316A10MM/1ML
Tebanicline dihydrochloride is a potent, orally effective analgesic that modulates neuronal nicotinic acetylcholine receptors (nAChRs). It exhibits high selectivity for neuronal nAChRs, acting as an agonist, and effectively inhibits cytisine binding to α4β2 neuronal nAChRs. This product is investigated for its potential in treating various types of pain and for its anxiolytic-like properties.
  • Potent and orally effective analgesic.
  • Highly selective for neuronal nAChRs.
  • Shows efficacy in models of acute, persistent, chemical, and neuropathic pain.
  • Investigated for anxiolytic-like activity.