Learn More
Medchemexpress LLC Tebanicline dihydrochloride | 209326-19-2 | 99.5% | C9H13Cl3N2O | 25 MG
SDP
Catalog No. 5000356853
Encompass_Preferred
Tebanicline dihydrochloride is a nAChR modulator with potent, orally effective analgesic activity. It inhibits the binding of cytisine to α4β2 neuronal nAChRs with a Ki of 37 pM. It is for research use only, and has shown antinociceptive effects in rat models of acute thermal, persistent chemical, and neuropathic pain.
  • Potent, orally effective analgesic activity
  • Inhibits binding of cytisine to α4β2 neuronal nAChRs with a Ki of 37 pM
  • Antinociceptive effects in rat models of acute thermal, persistent chemical, and neuropathic pain
  • Suitable for research use only
Catalog No. 50-003-56853 Supplier Medchemexpress LLC Supplier No. 25MGHY14316A
Tebanicline dihydrochloride is a nAChR modulator with potent, orally effective analgesic activity. It inhibits the binding of cytisine to α4β2 neuronal nAChRs with a Ki of 37 pM. It is for research use only, and has shown antinociceptive effects in rat models of acute thermal, persistent chemical, and neuropathic pain.
  • Potent, orally effective analgesic activity
  • Inhibits binding of cytisine to α4β2 neuronal nAChRs with a Ki of 37 pM
  • Antinociceptive effects in rat models of acute thermal, persistent chemical, and neuropathic pain
  • Suitable for research use only