Learn More
Medchemexpress LLC Tebanicline dihydrochloride | 209326-19-2 | 99.5% | C9H13Cl3N2O | 50 MG
SDP
Catalog No. 5000353839
Encompass_Preferred
Tebanicline dihydrochloride is a nAChR modulator with potent, orally effective analgesic activity. It inhibits the binding of cytisine to α4β2 neuronal nAChRs with a Ki of 37 pM.
  • nAChR modulator
  • Potent and orally effective analgesic activity
  • Inhibits binding of cytisine to α4β2 neuronal nAChRs with a Ki of 37 pM
  • Potent antinociceptive agent with full efficacy in models of acute and persistent pain
  • Produces significant antinociceptive effects in mice against acute noxious thermal stimulation
  • Orally active
  • Has antinociceptive effects in rat models of acute thermal, persistent chemical, and neuropathic pain
Catalog No. 50-003-53839 Supplier Medchemexpress LLC Supplier No. HY14316A50MG
Tebanicline dihydrochloride is a nAChR modulator with potent, orally effective analgesic activity. It inhibits the binding of cytisine to α4β2 neuronal nAChRs with a Ki of 37 pM.
  • nAChR modulator
  • Potent and orally effective analgesic activity
  • Inhibits binding of cytisine to α4β2 neuronal nAChRs with a Ki of 37 pM
  • Potent antinociceptive agent with full efficacy in models of acute and persistent pain
  • Produces significant antinociceptive effects in mice against acute noxious thermal stimulation
  • Orally active
  • Has antinociceptive effects in rat models of acute thermal, persistent chemical, and neuropathic pain