Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tebbe Reagent Solution

Catalog No. 380237 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
100 mL
4 x 25 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-023-7 4 x 25 mL
38-023-7100ML 100 mL
38-023-725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 38-023-7 Supplier Sigma Aldrich Supplier No. 3802374X25ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tebbe reagent is a Lewis acid stabilized organometallic compound, which is used to transform carbonyl groups into +a-substituted methylenes. It also reacts with a wide range of carbonyl compounds, including esters, amides, and lactones to yield corresponding olefins.Tebbe reagent can be used:For the conversion of carbonyl groups of chlorophyll derivatives into the corresponding exo-methylene (or vinylidene) groups.In the synthesis of +a-C-glycosides from 3-OH glycol esters.As a versatile methylenation reagent for the conversion of ketones and aldehydes to olefins. It offers facile reaction with hindered ketones and allows the conversion of esters to vinyl ethers.To olefinate aldehydes.To methylenate a chiral polyhydroxyketone with high diasteroselctivity.Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 67719-69-1
Density 0.927 g/mL at 25°C
Molecular Formula C13H18AlClTi
Linear Formula C13H18AlClTi
MDL Number MFCD00151575
Quantity 4 x 25 mL
Synonym Bis(cyclopentadienyl)-mu-chloro-f(dimethylaluminum)-mu-methylenetitanium
Molecular Weight (g/mol) 284.58 g/mol
Biological Activity Central Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.