Learn More
Medchemexpress LLC Tebipenem pivoxil | 161715-24-8 | 99.7% | 5 MG
SDP
Catalog No. 5000406659
Encompass_Preferred
Tebipenem pivoxil is an orally active antibiotic that works against various pathogenic bacteria by binding to penicillin-binding protein (PBP), thereby inhibiting cell wall synthesis. It is intended for research use only.
  • Orally active antibiotic
  • Binds penicillin-binding protein (PBP), thereby inhibiting cell wall synthesis
  • Excellent antibacterial activity against various pathogenic bacteria
  • Shows inhibition with MIC50s below 64 μg/mL against tested Gram-positive and Gram-negative bacteria
  • Significantly protects sepsis mice challenged with various pathogenic bacteria
  • Significantly increased the survival number of sepsis mice within a 168 h observation period
Catalog No. 50-004-06659 Supplier Medchemexpress LLC Supplier No. HYB03965MG
Tebipenem pivoxil is an orally active antibiotic that works against various pathogenic bacteria by binding to penicillin-binding protein (PBP), thereby inhibiting cell wall synthesis. It is intended for research use only.
  • Orally active antibiotic
  • Binds penicillin-binding protein (PBP), thereby inhibiting cell wall synthesis
  • Excellent antibacterial activity against various pathogenic bacteria
  • Shows inhibition with MIC50s below 64 μg/mL against tested Gram-positive and Gram-negative bacteria
  • Significantly protects sepsis mice challenged with various pathogenic bacteria
  • Significantly increased the survival number of sepsis mice within a 168 h observation period