Learn More
Cayman Chemical Tebipenem (hydrate), Purity: ≥95%, 100 mg
SDP
Catalog No. 502946754
Encompass_Preferred
A carbenapenem antibiotic; active against a panel of clinical isolates from a variety of bacterial species (MIC50s = ≤0.0039-8 µg/ml); inhibits β-lactamase; decreases the number of CFUs in the lungs in a mouse model of penicillin-resistant S. pneumoniae infection at doses ranging from 0.32 to 3.2 mg/kg
Catalog No. 50-294-6754 Supplier Cayman Chemical Supplier No. 33875100 MG
A carbenapenem antibiotic; active against a panel of clinical isolates from a variety of bacterial species (MIC50s = ≤0.0039-8 µg/ml); inhibits β-lactamase; decreases the number of CFUs in the lungs in a mouse model of penicillin-resistant S. pneumoniae infection at doses ranging from 0.32 to 3.2 mg/kg