Learn More
Crescent Chemical Co Inc TEBUCONAZOLE 250 MG
SDP
Catalog No. 5077061
Encompass_Preferred
Tebuconazole Fungicide (WS) CAS 107534-96-3 Molecular Weight - 307.8184 Storage - RT Catalog Num - DRE-C17178700 pesticide
Catalog No. 50-770-61 Supplier Crescent Chemical Co Inc Supplier No. PSC17178700
Tebuconazole Fungicide (WS) CAS 107534-96-3 Molecular Weight - 307.8184 Storage - RT Catalog Num - DRE-C17178700 pesticide