Learn More
Lkt Laboratories Tebuconazole, ≥98%, 5GR
SDP
Catalog No. 501534937
Encompass_Preferred
Triazole; 14-α demethylase inhibitor, voltage-gated Ca2+ channel blocker, potential aromatase inhibitor. CAS: 107534-96-3. Additional sizes available upon request. Ships from Saint Paul, MN.
Catalog No. 50-153-4937 Supplier Lkt Laboratories Supplier No. T16055G
Triazole; 14-α demethylase inhibitor, voltage-gated Ca2+ channel blocker, potential aromatase inhibitor. CAS: 107534-96-3. Additional sizes available upon request. Ships from Saint Paul, MN.