Learn More
Cayman Chemical TebufenozIde 100mg
SDP
Catalog No. 502933308
Encompass_Preferred
An insecticide and ecdysone receptor agonist (IC50 = 0.85 nM); induces a premature molt and is lethal to S. litura third instar larva (LD50 = 0.33 µM per larva); inhibits p-glycoprotein in LLC-PK1 porcine kidney epithelial cells transfected with MDR1 (IC50 = 21.5 µM); decreases colony formation and induces apoptosis and cell cycle arrest in HeLa cells when used at concentrations ranging from 50 to 200 UG/ml
Catalog No. 50-293-3308 Supplier Cayman Chemical Supplier No. 24055100 MG
An insecticide and ecdysone receptor agonist (IC50 = 0.85 nM); induces a premature molt and is lethal to S. litura third instar larva (LD50 = 0.33 µM per larva); inhibits p-glycoprotein in LLC-PK1 porcine kidney epithelial cells transfected with MDR1 (IC50 = 21.5 µM); decreases colony formation and induces apoptosis and cell cycle arrest in HeLa cells when used at concentrations ranging from 50 to 200 UG/ml