Learn More
Medchemexpress LLC HY-B2054 100mg Medchemexpress, Tebufenozide CAS:112410-23-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB2054100MG
Encompass_Preferred
Medchemexpress, HY-B2054 100mg Tebufenozide CAS:112410-23-8 Tebufenozide is a nonsteroidal ecdysone agonist used to control pest. Tebufenozide has cytotoxic and induces apoptosis in HeLa and insect Tn5B1-4 cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2721