Learn More
TARGETMOL CHEMICALS INC Tebufenozide 200MG
SDP
Catalog No. 502878542
Encompass_Preferred
Also available in 1 mL, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Tebufenozide is a novel nonsteroidal ecdysone agonist. It shows good efficacy and playing an increasingly important role in the control of Lepidopteran pests. Purity 98.04%
Catalog No. 50-287-8542 Supplier TARGETMOL CHEMICALS INC Supplier No. T19823200MG
Also available in 1 mL, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Tebufenozide is a novel nonsteroidal ecdysone agonist. It shows good efficacy and playing an increasingly important role in the control of Lepidopteran pests. Purity 98.04%