Learn More
Eurofins Discoverx Tec, activated, 250 µg. (14-801M)
Supplier:  Eurofins Discoverx 14801M
Encompass
N-terminal 6xHis-tagged recombinant, human Tec amino acids 174-end. Auto activated by incubating with Mg/ATP, excess ATP and MgAc removed by gel filtration. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9358