Learn More
Cayman Chemical Tecadenoson 100mg
SDP
Catalog No. 502938656
Encompass_Preferred
An adenosine A1 receptor agonist (Ki = 6.5 nM); selective for adenosine A1 over A2 receptors (Ki = 2,315 nM); prolongs SH intervals in isolated perfused guinea pig hearts (EC50 = 40.6 nM); induces PR interval prolongation in anesthetized guinea pigs (ED50 = 0.9 μg/kg); reduces heart rate in anesthetized rats
Catalog No. 50-293-8656 Supplier Cayman Chemical Supplier No. 28449100 MG
An adenosine A1 receptor agonist (Ki = 6.5 nM); selective for adenosine A1 over A2 receptors (Ki = 2,315 nM); prolongs SH intervals in isolated perfused guinea pig hearts (EC50 = 40.6 nM); induces PR interval prolongation in anesthetized guinea pigs (ED50 = 0.9 μg/kg); reduces heart rate in anesthetized rats