Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Tecaform Density Set
SCI_ED

Catalog No. S139420 Shop All Supertek Scientific Products
Change view
Click to view available options
Grade:
6+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S139420 6+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S139420 Supplier Supertek™ Supplier No. PH10048/4A
Add to Cart
Add to Cart
This item is not returnable. View return policy

The Tecaform Density Set demonstrates how density and specific gravity of each example remains the same, despite the mass and volume being different.

  • Consists of 12 tecaform cylinders of various lengths.
  • Cylinders stored in a wood storage block.

Specifications

Grade 6+
Format Set
Languages English
Subject Physcial Science - Density
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.