Learn More
Medchemexpress LLC HY-10167A 10mg Medchemexpress, Tecalcet (Hydrochloride) CAS:177172-49-5 Purity:>98%
SDP
Catalog No. 501967864
Encompass_Preferred
Medchemexpress, HY-10167A 10mg Tecalcet (HCl) CAS:177172-49-5 Tecalcet Hydrochloride (R 568 Hydrochloride), an orally active calcimimetic compound, allosterically and positively modulates the calcium-sensing receptor (CaSR). Tecalcet Hydrochloride (R 568 Hydrochloride) increases the sensitivity to activation by extracellular Ca2+[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7864 Supplier Medchemexpress LLC Supplier No. HY10167A10MG
Medchemexpress, HY-10167A 10mg Tecalcet (HCl) CAS:177172-49-5 Tecalcet Hydrochloride (R 568 Hydrochloride), an orally active calcimimetic compound, allosterically and positively modulates the calcium-sensing receptor (CaSR). Tecalcet Hydrochloride (R 568 Hydrochloride) increases the sensitivity to activation by extracellular Ca2+[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.