Learn More
Medchemexpress LLC HY-10167A 5mg Medchemexpress, Tecalcet (Hydrochloride) CAS:177172-49-5 Purity:>98%
SDP
Catalog No. 501932193
Encompass_Preferred
Medchemexpress, HY-10167A 5mg Tecalcet (Hydrochloride) CAS:177172-49-5 Tecalcet Hydrochloride (R 568 Hydrochloride), an orally active calcimimetic compound, allosterically and positively modulates the calcium-sensing receptor (CaSR) . Tecalcet Hydrochloride (R 568 Hydrochloride) increases the sensitivity to activation by extracellular Ca 2+ [1] [2] [3] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2193 Supplier Medchemexpress LLC Supplier No. HY10167A5MG
Medchemexpress, HY-10167A 5mg Tecalcet (Hydrochloride) CAS:177172-49-5 Tecalcet Hydrochloride (R 568 Hydrochloride), an orally active calcimimetic compound, allosterically and positively modulates the calcium-sensing receptor (CaSR) . Tecalcet Hydrochloride (R 568 Hydrochloride) increases the sensitivity to activation by extracellular Ca 2+ [1] [2] [3] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.