Learn More
Medchemexpress LLC Tecalcet hydrochloride | 177172-49-5 | MFCD16038895 | 99.7% | 340.29 g/mol | C18H23Cl2NO | 1 ML
SDP
Catalog No. 5000249102
Encompass_Preferred
Tecalcet hydrochloride is an orally active calcimimetic that allosterically and positively modulates the calcium-sensing receptor (CaSR). It is provided for research use in solution and solid forms and is commonly used in in vitro pharmacology and calcium-signaling studies.
  • Orally active calcimimetic that modulates the calcium-sensing receptor.
  • Suitable for in vitro pharmacology and CaSR signaling studies.
  • Available as a 10 mM solution in DMSO (1 mL) and as solid quantities for formulation flexibility.
  • High chemical purity (≈99.7%) for reliable experimental results.
  • Molecular weight 340.29 g/mol and molecular formula C18H23Cl2NO.
Catalog No. 50-002-49102 Supplier Medchemexpress LLC Supplier No. HY10167A10MM 1ML
Tecalcet hydrochloride is an orally active calcimimetic that allosterically and positively modulates the calcium-sensing receptor (CaSR). It is provided for research use in solution and solid forms and is commonly used in in vitro pharmacology and calcium-signaling studies.
  • Orally active calcimimetic that modulates the calcium-sensing receptor.
  • Suitable for in vitro pharmacology and CaSR signaling studies.
  • Available as a 10 mM solution in DMSO (1 mL) and as solid quantities for formulation flexibility.
  • High chemical purity (≈99.7%) for reliable experimental results.
  • Molecular weight 340.29 g/mol and molecular formula C18H23Cl2NO.