Learn More
Fluotics Tecan Fluent Evo LiHa Compatible P200, sterile
Catalog No. 5000435228
Encompass
Tecan Fluent Evo LiHa Compatible P200, sterile, Working Volume: 200uL, Tip Color: Clear, Hanging Rack, LiHA/FCA, Compare To: 30126017, 24 racks/case with 96 tips per rack
Catalog No. 50-004-35228 Supplier Fluotics Supplier No. LHA200.ST
Tecan Fluent Evo LiHa Compatible P200, sterile, Working Volume: 200uL, Tip Color: Clear, Hanging Rack, LiHA/FCA, Compare To: 30126017, 24 racks/case with 96 tips per rack