Learn More
Fluotics Tecan Evo/Fluent Compatible P200, filter/sterile
Catalog No. 5000435278
Encompass
Tecan Evo/Fluent Compatible P200, filter/sterile, Working Volume: 200uL, Tip Color: Black, Single Stack, base included, LiHa/FCA, Compare To: 30057821, 40 racks/case with 96 tips per rack
Catalog No. 50-004-35278 Supplier Fluotics Supplier No. LHAB200CBR.F
Tecan Evo/Fluent Compatible P200, filter/sterile, Working Volume: 200uL, Tip Color: Black, Single Stack, base included, LiHa/FCA, Compare To: 30057821, 40 racks/case with 96 tips per rack