Learn More
Fluotics Tecan Evo/Fluent Compatible P200, filter/sterile
Supplier: Fluotics LHAB200R.F
Tecan Evo/Fluent Compatible P200, filter/sterile, Working Volume: 200uL, Tip Color: Clear, Single Stack, base included, LiHa/FCA, 40 racks/case with 96 tips per rack