Learn More
Fluotics Tecan Fluent Evo MCA compatible P200, sterile
Catalog No. 5000435147
Encompass_Preferred
Tecan Fluent Evo MCA compatible P200, sterile, Working Volume: 200uL, Tip Color: Clear, Tip box, 96 MCA, Compare To: 30038617, 50 racks/case with 96 tips per rack
Catalog No. 50-004-35147 Supplier Fluotics Supplier No. MCA200XL.ST
Tecan Fluent Evo MCA compatible P200, sterile, Working Volume: 200uL, Tip Color: Clear, Tip box, 96 MCA, Compare To: 30038617, 50 racks/case with 96 tips per rack