Learn More
Fluotics Tecan Evo/Fluent Compatible P50, filter/sterile
Catalog No. 5000435169
Encompass
Tecan Evo/Fluent Compatible P50, filter/sterile, Working Volume: 50uL, Tip Color: Clear, Single Stack, base included, LiHa/FCA, 40 racks/case with 96 tips per rack
Catalog No. 50-004-35169 Supplier Fluotics Supplier No. LHAB50R.F
Tecan Evo/Fluent Compatible P50, filter/sterile, Working Volume: 50uL, Tip Color: Clear, Single Stack, base included, LiHa/FCA, 40 racks/case with 96 tips per rack