Learn More
Fluotics Tecan Fluent COMBI Compatible P200, filter/sterile
Catalog No. 5000435191
Encompass
Tecan Fluent COMBI Compatible P200, filter/sterile, Working Volume: 200uL, Tip Color: Black, Single Stack, COMBI MCA LiHa/FCA, 24 racks/case with 96 tips per rack
Catalog No. 50-004-35191 Supplier Fluotics Supplier No. CLHA200CBR.F
Tecan Fluent COMBI Compatible P200, filter/sterile, Working Volume: 200uL, Tip Color: Black, Single Stack, COMBI MCA LiHa/FCA, 24 racks/case with 96 tips per rack