Learn More
Fluotics Tecan Fluent COMBI Compatible P10, filter/sterile
Catalog No. 5000435256
Encompass
Tecan Fluent COMBI Compatible P10, filter/sterile, Working Volume: 10uL, Tip Color: Clear, Single Stack, COMBI MCA LiHa/FCA, 24 racks/case with 96 tips per rack
Catalog No. 50-004-35256 Supplier Fluotics Supplier No. CLHA10R.F
Tecan Fluent COMBI Compatible P10, filter/sterile, Working Volume: 10uL, Tip Color: Clear, Single Stack, COMBI MCA LiHa/FCA, 24 racks/case with 96 tips per rack