Learn More
Fluotics Tecan Fluent COMBI Compatible P50, filter/sterile
Catalog No. 5000435261
Encompass
Tecan Fluent COMBI Compatible P50, filter/sterile, Working Volume: 50uL, Tip Color: Clear, Single Stack, Tip box base included, COMBI MCA LiHa/FCA, 40 racks/case with 96 tips per rack
Catalog No. 50-004-35261 Supplier Fluotics Supplier No. CLHAB50R.F
Tecan Fluent COMBI Compatible P50, filter/sterile, Working Volume: 50uL, Tip Color: Clear, Single Stack, Tip box base included, COMBI MCA LiHa/FCA, 40 racks/case with 96 tips per rack