Learn More
Fluotics Tecan Fluent COMBI Compatible P200, filter/sterile
Supplier:  Fluotics CLHA200R.F
Encompass
Tecan Fluent COMBI Compatible P200, filter/sterile, Working Volume: 200uL, Tip Color: Clear, Single Stack, COMBI MCA LiHa/FCA, 24 racks/case with 96 tips per rack
Catalog No. 50-004-35119