Learn More
Fluotics Tecan Evo/Fluent Compatible P10, filter/sterile
Supplier:  Fluotics LHA10CB.F
Encompass
Tecan Evo/Fluent Compatible P10, filter/sterile, Working Volume: 10uL, Tip Color: Black, Hanging Rack, LiHa/FCA, Compare To: 30104804, 24 racks/case with 96 tips per rack
Catalog No. 50-004-35125