Learn More
Fluotics Tecan Fluent Evo LiHa Compatible P200, filter/sterile
Catalog No. 5000435211
Encompass
Tecan Fluent Evo LiHa Compatible P200, filter/sterile, Working Volume: 200uL, Tip Color: Black, Hanging Rack, LiHA/FCA, Compare To: 30184272, 30000629, 24 racks/case with 96 tips per rack
Catalog No. 50-004-35211 Supplier Fluotics Supplier No. LHA200CB.F
Tecan Fluent Evo LiHa Compatible P200, filter/sterile, Working Volume: 200uL, Tip Color: Black, Hanging Rack, LiHA/FCA, Compare To: 30184272, 30000629, 24 racks/case with 96 tips per rack