Learn More
Fluotics Tecan Fluent Evo LiHa Compatible P50, filter/sterile
Catalog No. 5000435214
Encompass
Tecan Fluent Evo LiHa Compatible P50, filter/sterile, Working Volume: 50uL, Tip Color: Black, Hanging Rack, LiHA/FCA, Compare To: 30032114, 24 racks/case with 96 tips per rack
Catalog No. 50-004-35214 Supplier Fluotics Supplier No. LHA50CB.F
Tecan Fluent Evo LiHa Compatible P50, filter/sterile, Working Volume: 50uL, Tip Color: Black, Hanging Rack, LiHA/FCA, Compare To: 30032114, 24 racks/case with 96 tips per rack