Learn More
Fluotics Tecan Fluent Evo MCA compatible P50, sterile
Catalog No. 5000435170
Encompass_Preferred
Tecan Fluent Evo MCA compatible P50, sterile, Working Volume: 50uL, Tip Color: Clear, Tip box, 96 MCA, Compare To: 30038607, 50 racks/case with 96 tips per rack
Catalog No. 50-004-35170 Supplier Fluotics Supplier No. MCA50.ST
Tecan Fluent Evo MCA compatible P50, sterile, Working Volume: 50uL, Tip Color: Clear, Tip box, 96 MCA, Compare To: 30038607, 50 racks/case with 96 tips per rack