Learn More
Fluotics Tecan Fluent Evo MCA compatible P500, sterile
Catalog No. 5000435204
Encompass
Tecan Fluent Evo MCA compatible P500, sterile, Working Volume: 500uL, Tip Color: Clear, Tip box, 96 MCA, Compare To: 30038607, 50 racks/case with 96 tips per rack
Catalog No. 50-004-35204 Supplier Fluotics Supplier No. MCA500.ST
Tecan Fluent Evo MCA compatible P500, sterile, Working Volume: 500uL, Tip Color: Clear, Tip box, 96 MCA, Compare To: 30038607, 50 racks/case with 96 tips per rack