Learn More
Medchemexpress LLC HY-14854 5mg Medchemexpress, Tecarfarin CAS:867257-26-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY148545MG
Encompass_Preferred
Medchemexpress, HY-14854 5mg Tecarfarin CAS:867257-26-9 Tecarfarin (ATI-5923) is an orally active and non-competitive vitamin K epoxide reductase (VKOR) antagonist, and impairs the activation of the vitamin K-dependent clotting factors II, VII, IX and X. Tecarfarin has the antithrombotic activity . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9919