Learn More
TARGETMOL CHEMICALS INC TECASTEMIZOLE 5MG
SDP
Catalog No. 502580712
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tecastemizole (R 43512) is a selective antagonist of H1 receptor and a major metabolite of astemizole with anti-inflammatory effects. purity: 99%
Catalog No. 50-258-0712 Supplier TARGETMOL CHEMICALS INC Supplier No. T262535MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tecastemizole (R 43512) is a selective antagonist of H1 receptor and a major metabolite of astemizole with anti-inflammatory effects. purity: 99%