Learn More
Description
- Reliable All-rounders
- High quality buffers - These solutions are traceable to the primary standards of reference materials selected by NIST.
- Quality inspection certificate included - For record and archiving purposes
- SDS easily available - All Safety Data Sheets have been created in accordance with GHS (Globally Harmonized System).
Specifications
Specifications
| Color | Colorless |
| Description | pH Buffer |
| For Use With (Application) | Calibration |
| For Use With (Equipment) | pH Meter and Electrode |
| pH | 2 |
| Physical Form | Liquid |
| Quantity | 30 Ea. |
| Solution Type | Calibration |
| Grade | Not Available |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
