Learn More
Cole-Parmer Dri-Block Aluminum Block Insert, 20 x 13 mm dia; 48-mm deep, conical well
Catalog No. NC0758465 Shop All Cole-Parmer Products
Encompass_Preferred
Techne Dri-Block Aluminum Heating Block Insert, 20 x 13 mm diameter, Precision-engineered aluminum heating blocks offer excellent temperature accuracyVersatile range of interchangeable heating blocks work with many size sample tubes and platesHeaters sold separately
Catalog No. NC0758465 Supplier Cole-Parmer Supplier No. 3662054
Techne Dri-Block Aluminum Heating Block Insert, 20 x 13 mm diameter, Precision-engineered aluminum heating blocks offer excellent temperature accuracyVersatile range of interchangeable heating blocks work with many size sample tubes and platesHeaters sold separately