Learn More
Cole-Parmer Techne IFB-52 Industrial Fluidized Sand Bath, 12 L
Catalog No. NC1486486 Shop All Cole-Parmer Products
Encompass
Safe, efficient and cost-effective cleaning of tooling, components, and parts. Removes plastics, paints, epoxy, rubber, adhesives, grease, lubricants, and oils. Fast cleaning minimizes production equipment down-time. No tool damage or abrasion in cleaning. Dry, inert, nontoxic aluminum oxide is a safer alternative than dangerous chemicals or solvents. Sand sold separately.
Catalog No. NC1486486 Supplier Cole-Parmer Supplier No. 0118420
Add to Cart
Add to Cart
Request a Quote
Safe, efficient and cost-effective cleaning of tooling, components, and parts. Removes plastics, paints, epoxy, rubber, adhesives, grease, lubricants, and oils. Fast cleaning minimizes production equipment down-time. No tool damage or abrasion in cleaning. Dry, inert, nontoxic aluminum oxide is a safer alternative than dangerous chemicals or solvents. Sand sold separately.