Learn More
Azzota Corp Technical Balances-10000, Technical Balances, Readability: 0.1G, Weighting Mode:Single, Capacity/Range:10000G, Linearity: 0.1G, Pan Size: 195 X 175 mm
Catalog No. 501613021
Encompass_Preferred
Technical Balances-10000, Technical Balances, Readability: 0.1g, Weighting Mode:Single, Capacity/Range:10000g, Linearity: 0.1g, Pan size: 195 x 175 mm
Catalog No. 50-161-3021 Supplier Azzota Corp Supplier No. TSM10000
Technical Balances-10000, Technical Balances, Readability: 0.1g, Weighting Mode:Single, Capacity/Range:10000g, Linearity: 0.1g, Pan size: 195 x 175 mm