Learn More
Description
- It ensures accurate and reliable measurements in highly acidic environments, making it ideal for laboratories, industrial processes, and research.
- The 250 mL size supports multiple calibrations, aiding consistent pH accuracy and regulatory compliance.
Specifications
Specifications
| For Use With (Application) | Calibration |
| For Use With (Equipment) | pH Meter and Electrode |
| pH | 1.00 |
| Physical Form | Solution |
| Quantity | 250 mL |
| Solution Type | Technical Buffer |
| Grade | Laboratory |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.