Learn More
Aqua Solutions Technicon Standard #1 0.25 gpL NaCL0₃, 100 gpL NaCl, 200 gpL NaOH (1L)
SDP
Catalog No. NC9524037
Encompass_Preferred
Technicon Standard #1 0.25 gpL NaCL0₃, 100 gpL NaCl, 200 gpL NaOH (1L)
Catalog No. NC9524037 Supplier Aqua Solutions Supplier No. VUL363
Technicon Standard #1 0.25 gpL NaCL0₃, 100 gpL NaCl, 200 gpL NaOH (1L)